Learn More
JDM SCIENTIFIC LLC AlteCap™ Switch Cassette 12/96 - Internal thread Matrix/Nunc Bank-It vials
Supplier:  JDM SCIENTIFIC LLC 404027
Encompass_Preferred
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Matrix/Nunc Bank-It vials - part numbers 3748-BR, 374078, 374074
Catalog No. 50-271-9987
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.