Learn More
JDM SCIENTIFIC LLC AlteRead Wireless Single tube 2D & 1D barcode camera reader
Single tube 2D & 1D barcode wireless camera reader. Compatible with all 2D and 1D tubes. Scans 2D and 1D barcodes directly to any application. Can be moved easily around the lab and used in hoods. Compatible with IOS, Windows and mobile phone operating systems. Warranty: 5 years