Learn More
Crescent Chemical Co Inc ALU-GEL-S SUSPENSION STERILE
SDP
Supplier:  Crescent Chemical Co Inc SE12261.02
Encompass_Preferred
Alu-Gel-S suspension research grade sterile Ph. Eur. 1.3 % in water. Pyrogen free (as assayed in the supernatant), aged, salt-free. Size - 10x50ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 12261.02
Catalog No. 50-247-278
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.