Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder, Approx. 30%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
125 g
500 g
2.5 kg
150 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder is a salt that is used as a topical astringent to treat skin irritations from poison ivy, substances such as soaps and cosmetics and insect bites. Aluminum acetate is particularly helpful for foot problems including athlete’s foot, swelling, and excessive sweating. It is also used to treat ear canal infections. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS Max % | 30%, 70% |
Quantity | 125 g |
Percent Purity | ∼30% |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction