Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Aluminum Ammonium Sulfate Dodecahydrate
SCI_ED

Catalog No. S25142A Shop All AquaPhoenix Scientific Products
Click to view available options
:

Specifications

Color

White

CAS

7784-26-1

Physical Form

Solid

Quantity

500 g

Molecular Formula

AlH28NO20S2

Linear Formula

AlNH4(SO4)2·12H2O

Synonym

ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate

InChI Key

WZUKKIPWIPZMAS-UHFFFAOYSA-K

SMILES

[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]

IUPAC Name

aluminum;azanium;disulfate;dodecahydrate

Molecular Weight (g/mol)

453.313

PubChem CID

62668

Formula Weight

453.32

Grade

Reagent

Packaging

Reagent Grade

Density

1.6g/mL

Chemical Name or Material

Aluminum Ammonium Sulfate, Dodecahydrate

ShelfLife

5 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.