Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Aluminum Oxide, Anhydrous
SCI_ED

Catalog No. p-4416319 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade - Powder

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Solid
Quantity 500 g
Linear Formula Al2O3
Formula Weight 101.96
Packaging Lab Grade - Powder
Density 4g/mL
Chemical Name or Material Aluminum Oxide, Anhydrous
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.