Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Aluminum Potassium Sulfate, Dodecahydrate
SCI_ED

Catalog No. S25152 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

7784-24-9

Molecular Formula

AlH24KO20S2

Molecular Weight (g/mol)

474.372

InChI Key

GNHOJBNSNUXZQA-UHFFFAOYSA-J

Synonym

kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate

PubChem CID

62667

ChEBI

CHEBI:86465

IUPAC Name

aluminum;potassium;disulfate;dodecahydrate

SMILES

O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Color

White

Quantity

30 g

Linear Formula

AlK(SO4)2 ·12H2O

Formula Weight

474.3558

Packaging

Lab Grade - Powder

Chemical Name or Material

Aluminum Potassium Sulfate, Dodecahydrate

ShelfLife

5 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.