Learn More
Glass Expansion Aluminum Sampler
Supplier: Glass Expansion TF1001AL
Aluminum Sampler Cone – For Thermo/Finnigan Element 1, 2 & Neptune ICP-MS. Use for lowest Ni background, and laser ablation applications. Not recommended for samples containing hydrofluoric acid (HF). Use a new Graphite Sampler Cone Gasket (501907717) each time the cone is re-installed, even if it’s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background and semiconductor applications, we recommend a platinum cone, see catalog number 501907711.For high sensitivity applications and low matrix containing samples, we recommend Nickel Jet Sampler, see catalog number 501907709.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.