Learn More
Medchemexpress LLC HY-101253 5mg Medchemexpress, AM-92016 (hydrochloride) CAS:133229-11-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012535MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-101253 5mg AM-92016 (hydrochloride) CAS:133229-11-5 AM-92016 hydrochloride is a specific blocker of rectifier potassium current ( IK ). AM-92016 hydrochloride delays rectifier potassium channel (IK), repolarizes the membrane thereby restricting the duration of the nerve impulse thereby restricting the duration of the nerve impulse [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2076
missing translation for 'QuantityLeft'