Learn More
TARGETMOL CHEMICALS INC AM966 25MG
Supplier:  TARGETMOL CHEMICALS INC T4278
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AM966 a excellent affinity specific oral LPA1(IC50=17 nM) antagonist suppresses LPA-stimulated intracellular calcium release. purity: 98%
Catalog No. 50-257-9915
missing translation for 'QuantityLeft'