Learn More
Accuflow Systems Accuflow Systems Sterile Empty Vial (SEV), 2.0 cc, 13 mm glass with silver cap - 25/pack
ALK Sterile Empty Vials (SEV) feature Type 1 glass USP borosilicate, and a latex free stopper. ALK SEVs are produced at a cGMP FDA Manufacturer and USP tested for sterility and endotoxins. Certificate of Analysis available for all production lots and vials are aseptically assembled in an ISO 5 (class 100) cleanroom. 25/PACK