Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMD 3465 hexahydrobromide, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective CXCR4 antagonist; exhibits 8-fold higher affinity than AMD 3100 (Cat.No. 3299). Inhibits SDF-1α-ligand binding (Ki = 41.7 nM). Potently inhibits HIV cell entry in vitro; causes leukocytosis and mobilizes haematopoietic stem cells in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 896.07 |
Percent Purity | >97% |
Chemical Name or Material | AMD 3465 hexahydrobromide |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction