Learn More
Electron Microscopy Sciences AMD-SS Balance Table W/Isolation Area, 75 x 75 cm, Stainless Steel
Supplier: Electron Microscopy Sciences 6690

This Stainless balance workstation features an AMS-30x45 platform with four 7Hz sorbothane isolators which give an excellent 95 percent isolation at 50Hz. Features a very low Ra of 0.4, making it ideal for cleanrooms as well as being resistant to almost all forms of biological and chemical attack. A unique sound deadening core soaks up surface borne vibrations and ensures our tables outerperform simple granite and steel tables by up to 20dB. 240 silicon satin stainless ensures our tables meet all the requirements for use in stringent Steel Case environments such as cleanrooms and biohazard areas. Four 7Hz sorbothane isolators give an excellent 95 percent isolation at 50Hz.
Size: 75 x 75 cm
Type: Stainless Steel
Surface: 4 Sorbothane
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.