Learn More
Enzo Life Sciences AMH (human) (recombinant) (10µg)
Alternative name: anti-Müllerian hormone, Mullerian-inhibiting substance. Concentration: 0.1mg/ml when reconstituted as suggested. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized in 1X PBS, pH 7.4. Source: Produced in HEK 293 cells. AMH (aa 19-560) is fused at the N-terminus to a FLAG-tag and His-tag. Long Term Storage: -20°C.