Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Catalog No. AC291640050
missing translation for 'orderingAttributeHoverText'
Quantity:
Packaging:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

1066-51-9

Molecular Formula

CH6NO3P

Molecular Weight (g/mol)

111.04

MDL Number

MFCD00008105

InChI Key

MGRVRXRGTBOSHW-UHFFFAOYSA-N

Synonym

aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl

PubChem CID

14017

ChEBI

CHEBI:28812

IUPAC Name

aminomethylphosphonic acid

SMILES

C(N)P(=O)(O)O

Specifications

CAS Min %

98.5

CAS Max %

100.0

Melting Point

>300.0°C

Color

White

Infrared Spectrum

Authentic

Assay Percent Range

99%

Packaging

Glass bottle

Linear Formula

H2NCH2P(O)(OH)2

Quantity

5 g

Beilstein

01, 2596

Solubility Information

Solubility in water: practically insoluble. Other solubilities: soluble in alcohols

Formula Weight

111.04

Percent Purity

99%

Physical Form

Fine Crystalline Powder

Chemical Name or Material

(Aminomethyl)phosphonic acid

missing translation for 'healthHazard1'

GHS Signal Word: Danger

missing translation for 'healthHazard2'

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

missing translation for 'healthHazard3'

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.