Learn More
Medchemexpress LLC HY-14545 100mg Medchemexpress, Amisulpride CAS:71675-85-9 Purity:>98%
Supplier: Medchemexpress LLC HY14545100MG
Medchemexpress, HY-14545 100mg Amisulpride CAS:71675-85-9 Amisulpride is a dopamine D2/D3 receptor antagonist with Kis of 2.8 and 3.2 nM for human dopamine D2 and D3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.