Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amlexanox, Tocris Bioscience™

Catalog No. p-4816535 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of TBK

Selective inhibitor of TANK-binding kinase 1 (TBK1) and IKKε (IC50 values are ∽1-2 μM). Displays no effect on IKKα or IKKβ at these concentrations. Reversibly lowers weight, increases insulin sensitivity, and reduces inflammation and steatosis in three mouse models of obesity. Exhibits antiallergic activity; inhibits the release of histamine from rat mast cells. Also binds to Hsp90 and inhibits C-terminal chaperone activity in vitro.

Chemical Identifiers

CAS 68302-57-8
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.30
MDL Number MFCD00864790
InChI Key SGRYPYWGNKJSDL-UHFFFAOYSA-N
Synonym amlexanox, aphthasol, amoxanox, 2-amino-7-isopropyl-5-oxo-5h-chromeno 2,3-b pyridine-3-carboxylic acid, amlenanox, solfa, amlexanoxo, amlexanoxum, amlexanoxum latin, amlexanoxo spanish
PubChem CID 2161
ChEBI CHEBI:31205
IUPAC Name 2-amino-5-oxo-7-(propan-2-yl)-5H-chromeno[2,3-b]pyridine-3-carboxylic acid
SMILES CC(C)C1=CC=C2OC3=NC(N)=C(C=C3C(=O)C2=C1)C(O)=O

Specifications

Quantity 10 mg
Formula Weight 298.29
Percent Purity >99%
Chemical Name or Material Amlexanox
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.