Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amlodipine Besylate, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. p-7156983 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

A3950, 111470-99-6, C20H25ClN2O5•C6H6O3S

Spectrum™ Chemical Amlodipine Besylate, USP is a medication used to treat high blood pressure and coronary artery disease. It is on the World Health Organization′s List of Essential Medicines. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 111470-99-6
Molecular Formula C26H31ClN2O8S
Molecular Weight (g/mol) 567.05
InChI Key ZPBWCRDSRKPIDG-UHFFFAOYNA-N
IUPAC Name 3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate; benzenesulfonic acid
SMILES OS(=O)(=O)C1=CC=CC=C1.CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Packaging Amber Glass Bottle
Quantity 5 g
Optical Rotation -0.10° to +0.10°
Percent Purity 97 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.