Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia-Ammonium Chloride Buffer, T.S., BAKER ANALYZED™ Reagent, J.T.Baker™

Catalog No. 02002264 Shop All Avantor Products
Click to view available options
Quantity:
1L
Packaging:
Narrow Mouth Polyethylene Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

Chemical Name or Material Ammonia-Ammonium Chloride Buffer, T.S.

Specifications

Packaging Narrow Mouth Polyethylene Bottle
Quantity 1L
Grade BAKER ANALYZED™ Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.