Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate, puriss., Honeywell Fluka™

Catalog No. 6001898 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:

Puriss., meets analytical specification of NF, Ph. Franc., FCC

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS

506-87-6

Molecular Formula

CH8N2O3

Molecular Weight (g/mol)

96.086

MDL Number

MFCD00010890

InChI Key

PRKQVKDSMLBJBJ-UHFFFAOYSA-N

Synonym

ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate

PubChem CID

517111

IUPAC Name

diazanium;carbonate

SMILES

C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Quantity

2.5 kg

Linear Formula

(NH4)2CO3

Beilstein

4253487

Formula Weight

96.09g/mol

Grade

puriss.

Residue after Ignition

≤0.05% (as SO4)

Density

Vapor Density: 2.7 (vs air)

Chemical Name or Material

Ammonium Carbonate, Meets analytical specification of NF, Ph.Franc.,FCC

Hazard Statement

P301 + P312 + P330

Precautionary Statement

H302

EINECSNumber

208-058-0

RTECSNumber

BP1925000

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.