Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Chloride
SCI_ED

Catalog No. S25168A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Freezing point depression

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

12125-02-9

Molecular Formula

ClH4N

Molecular Weight (g/mol)

53.49

MDL Number

MFCD00011420

InChI Key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Synonym

ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid

PubChem CID

25517

ChEBI

CHEBI:31206

IUPAC Name

amine hydrochloride

SMILES

N.Cl

Specifications

Color

White

Quantity

500 g

Assay Percent Range

100% w/v

Linear Formula

NH4Cl

Formula Weight

53.49

Grade

Laboratory

Packaging

Reagent Grade

Chemical Name or Material

Ammonium Chloride

ShelfLife

5 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.