Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Chloride ACS AR,Crystal, Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
Specifications
Specifications
Physical Form | Crystalline |
Quantity | 500 g |
Formula Weight | 53.49 |
Grade | ACS |
Packaging | POLYSTORMOR |
Chemical Name or Material | Ammonium Chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction