Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Dichromate
SCI_ED

Catalog No. S25169A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

7789-09-5

Molecular Formula

Cr2H8N2O7

Molecular Weight (g/mol)

252.06

InChI Key

JOSWYUNQBRPBDN-UHFFFAOYSA-P

Synonym

ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481

PubChem CID

24600

IUPAC Name

diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium

SMILES

[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Color

Orange to Red

Quantity

500 g

Assay Percent Range

99%

Linear Formula

(NH4)2Cr2O7

Formula Weight

252.06

Grade

Laboratory

Packaging

Lab Grade - Crystal

Density

2.15g/mL

Chemical Name or Material

Ammonium Dichromate

ShelfLife

1 year

DOTInformation

DOT Class 5.1, II, : Oxidizer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.