Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate (Granular/Certified ACS), Fisher Chemical™
GSA_VA

Catalog No. p-30976
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle

Decomposes at 210°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210°C
Melting Point 169°C
Color White
pH 4.5 to 6.0
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98.0 %
Linear Formula NH4NO3
UN Number UN 1942
Formula Weight 80.04
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 1.720g/cm³
Chemical Name or Material Ammonium Nitrate
Show More Show Less
DOTInformation DOT Class 5.1, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.