Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Oxalate, Monohydrate
SCI_ED

Catalog No. S25174A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

6009-70-7

Molecular Formula

C2H10N2O5

Molecular Weight (g/mol)

142.111

InChI Key

MSMNVXKYCPHLLN-UHFFFAOYSA-N

Synonym

ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate

PubChem CID

516808

IUPAC Name

diazanium;oxalate;hydrate

SMILES

C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Color

White

Quantity

500 g

Assay Percent Range

>99% w/v

Linear Formula

(NH4)2C2O4·H2O

Formula Weight

142.11

Grade

Laboratory

Packaging

Lab Grade - Crystal

Density

1.5g/mL

Chemical Name or Material

Ammonium Oxalate, Monohydrate

ShelfLife

5 years

DOTInformation

DOT Class 6.1, III, : Poison

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.