Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, ACS Reagent (Diammonium Hydrogen Phosphate), Reagents

Catalog No. 01338083
Click to view available options
Quantity:

Ammonium Phosphate Dibasic, ACS Reagent (Diammonium Hydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS

7783-28-0

Molecular Formula

H9N2O4P

Molecular Weight (g/mol)

132.06

MDL Number

MFCD00010891

InChI Key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Synonym

Diammonium Hydrogen Phosphate

IUPAC Name

phosphoric acid diamine

SMILES

N.N.OP(O)(O)=O

Specifications

Melting Point

155°C

Color

Colorless/White

Physical Form

Solid

Quantity

500 g

Grade

ACS

Packaging

Glass Bottle

Specific Gravity

1.619

Chemical Name or Material

Ammonium Phosphate Dibasic, ACS Reagent (Diammonium Hydrogen Phosphate)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.