Learn More
Bioworld Ammonium Phosphate, Monobasic, 500 g
Supplier:  Bioworld 401002121
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Itis formed when a solution of phosphoric acid is added to ammonia until the solution is distinctly acidic. It is often used in the blending of dry agricultural fertilizers. It supplies soil with the elements nitrogen and phosphorus in a form which is usable by plants. The compound is also a component of the ABC powder in some dry chemical fire extinguishers. It is a widely used crystal in the field of optics due to its birefringence properties. Commonly used in plant tissue culture media.ACS GradePrecipitate (in NH4OH): <0.005%Chloride: <0.0005%Nitrate: <0.001%Sulfur compounds: <0.005%Heavy metals (as Pb): <0.0005%Iron: 0.001%Potassium: <0.005%Sodium: <0.005%Appearance: White Crystalline PowderDensity : 1.80 g/cm3HS Code : 31054000IUPAC_Name: Ammonium Dihydrogen PhosphateMelting Point: 190 deg CPH Value: pH (5%, water, 25 C): 3.8-4.4RTECS Number: TC6587000
Catalog No. 50-255-271