Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Phosphate, Monobasic
SCI_ED

Catalog No. S25153A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

7722-76-1

Molecular Formula

H6NO4P

Molecular Weight (g/mol)

115.03

MDL Number

MFCD00003396

InChI Key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Synonym

ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate

PubChem CID

24402

ChEBI

CHEBI:62982

IUPAC Name

phosphoric acid amine

SMILES

N.OP(O)(O)=O

Specifications

Color

White

Quantity

500 g

Assay Percent Range

>98%

Linear Formula

NH4H2PO4

Formula Weight

115.03

Grade

Laboratory

Packaging

Lab Grade - Powder

Density

1.800

Chemical Name or Material

Ammonium Phosphate, Monobasic

ShelfLife

5 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.