Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Sulfate
SCI_ED

Catalog No. S25176A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

7783-20-2

Molecular Formula

H8N2O4S

Molecular Weight (g/mol)

132.13

MDL Number

MFCD00003391

InChI Key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Synonym

ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate

PubChem CID

6097028

ChEBI

CHEBI:62946

IUPAC Name

sulfuric acid diamine

SMILES

N.N.OS(O)(=O)=O

Specifications

Color

White

Quantity

500 g

Assay Percent Range

>95% w/v

Linear Formula

(NH4)2SO4

Formula Weight

132.14

Packaging

Reagent Grade

Density

1.8g/mL

Chemical Name or Material

Ammonium Sulfate

ShelfLife

5 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.