Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Thiocyanate
SCI_ED

Catalog No. S25158 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
Analytical reagent

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

1762-95-4

Molecular Formula

CH4N2S

Molecular Weight (g/mol)

76.117

InChI Key

SOIFLUNRINLCBN-UHFFFAOYSA-N

Synonym

ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt

PubChem CID

15666

IUPAC Name

azanium;thiocyanate

SMILES

C(#N)[S-].[NH4+]

Specifications

Color

White

Quantity

500 g

Assay Percent Range

100% w/w

Linear Formula

NH4SCN

Formula Weight

76.12

Grade

Reagent

Packaging

Lab Grade - Crystalline Powder

Density

1.3 g/cm3

Chemical Name or Material

Ammonium Thiocyanate

ShelfLife

2 years

DOTInformation

DOT Class 9, III, : Miscellaneous

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.