Learn More
Gold Biotechnology Inc Ampicillin (Sodium), USP Grade 5 g
SDP
Supplier:  Gold Biotechnology Inc A3015
Encompass
Ampicillin sodium is a member of the extended-spectrum β-lactam family and similar in structure to penicillin. It is used in the selection of antibiotic cell hybrids and has a high rate of action against many gram-negative bacterial strains. Ampicillin sodium targets non-ESBL (Extended Spectrum β-lactamase) bacteria including Staphylococcus and Streptococcus species and medically important enteric pathogens such as Shigella and Salmonella. It has been found to be effective against certain β-lactam sensitive VRE or vancomycin resistant Enterococcus species. Resistance to ampicillin is routinely utilized as a selectable marker to confirm successful cell transformation. Ampicillin Sodium is freely soluble in water.
Catalog No. NC2941520
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.