Learn More
Electron Microscopy Sciences Ampoule Cracker for 1-4 ml
Supplier:  Electron Microscopy Sciences 6060504
Encompass_Preferred
The Ampoule Cracker is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules. Simply slip the Ampoule Cracker over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.
Use: 1-4 ml
Catalog No. 50-997-516
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.