Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMT hydrochloride, Tocris Bioscience™

Catalog No. p-4783252 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective iNOS inhibitor

Potent, selective and reversible inhibitor of iNOS (IC50 = 3.6 nM; approximately 30 and 40 times selective over nNOS and eNOS respectively).

Chemical Identifiers

CAS 21463-31-0
Molecular Formula C5H10N2S
Molecular Weight (g/mol) 130.21
MDL Number MFCD00717539
InChI Key BATVOUKHGLKDGQ-SCSAIBSYSA-N
Synonym amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine, +/--amt hydrochloride, 2-amino-5,6-dihydro-6-methyl-4h-1,3-thiazine hydrochloride, 6-methyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, y-amt hydrochloride, ?-amt hydrochloride, c5h10n2s.hcl, inverted question mark-amt hydrochloride
PubChem CID 2733501
IUPAC Name (6R)-6-methyl-5,6-dihydro-4H-1,3-thiazin-2-amine
SMILES C[C@@H]1CCN=C(N)S1

Specifications

Quantity 10 mg
Formula Weight 166.67
Percent Purity >98%
Chemical Name or Material AMT hydrochloride
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.