Learn More
New England Biolabs, Inc. AMV Reverse Transcriptase - 200 units
SDP
Supplier:  New England Biolabs, Inc. M0277S
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
AMV Reverse Transcriptase - 200 units. Avian Myeloblastosis Virus (AMV) Reverse Transcriptase is an RNA-directed DNA polymerase. This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template.Help NEB go green. FREE FREIGHT on NEB orders greater than $350.00!
Catalog No. 50-995-213
missing translation for 'QuantityLeft'