Learn More
Bachem (Des-Gly10,D-Arg6,Pro-NHEt9)-LHRH (salmon) Acetate
SDP
Supplier:  Bachem 4016385.0025
Encompass_Preferred
(Des-Gly10,D-Arg6,Pro-NHEt9)-LHRH (salmon) Acetate, Pyr-His-Trp-Ser-Tyr-D-Arg-Trp-Leu-Pro-NHEt, 96497-82-4, C64H83N17O12, Mr 1282.47, 25mg. sGnRH-A. Analog of salmon GnRH showing a very high affinity to GnRH receptors used in aquaculture. Usually applied in combination with a dopamine antagonist as domperidone or pimozide, sGnRHa induces ovulation and spawning in a large number of fish species.
Catalog No. 50-194-6989
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.