Learn More
Andwin Scientific Sterile Urine Cup with Boritex™ Preservative
- Sterile 90mL container
- Boritex boric acid tablet keeps urine stable for up to 96 hours
- Ideal preservative system for remote collection sites and delayed laboratory examination
- Maintains formed elements in urine sediment and key diagnostic markers
- Provided with patient identification label
Specifications
Product Type
Urine Collection Cup
Closure Color
Blue
Closure Type
Screw Cap
Material
Polypropylene
Sterility
Non-sterile
Color
Clear
For Use With (Application)
For tests and studies, such as Urine chemistry or cultures
Includes
200mg Boritex Boric Acid tablet, 90 mL 53 mm Container and cap
Volume (English)
30.4 oz.
Volume (Metric)
90 mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.