Learn More
World Precision Instrument Anis Needle Holder without Lock, 11.5 cm, Curved, 0.5 mm Extra Delicate Smooth Tip, Round Handle
SDP
Supplier:  World Precision Instrument WP6037
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Anis needle holders are surgical instruments designed for holding and manipulating needles during suturing procedures. These needle holders feature knurled round handles for a secure grip, facilitating precise control during surgery. The knurled texture enhances maneuverability, especially in delicate and fine tissue suturing. The 12 cm (4.7") version with a lock incorporates a mechanism that allows you to secure the needle in place, providing stability during the suturing process. This locking feature ensures that the needle remains firmly held, preventing unintentional slippage and contributing to the accuracy of the suturing. The 11.5 cm (4.5") version without a lock lack this mechanism, offering a more dynamic handling experience.
Catalog No. 50-253-8281
missing translation for 'QuantityLeft'