Learn More
Invitrogen™ Annexin V Ready Flow Conjugates for Apoptosis Detection

Enjoy the convenience of ready-to-use Annexin V Ready Flow stand-alone conjugate stains, which offer up to 100-fold fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry. There is no need to dilute, weigh, or pipette the conjugate stains; simply add one drop into cells, incubate, and analyze.
The Annexin V Ready Flow conjugates are provided as ready-to-use, direct-to-sample apoptotic cell stains. Simply add one drop from the dropper bottle directly into 1 x 106 cells in 5 mM Ca2+-containing cell culture medium or annexin binding buffer (Cat. Nos. V13246 or BMS500BB), incubate, and analyze. The traditional annexin staining protocol can be used as an alternative method by replacing the addition of 5 μL annexin conjugate with the addition of 1 drop from the Ready Flow dropper bottle.
The Annexin V Alexa Fluor 488 conjugate is excitable at 495 nm with an emission peak at 519 nm.
The Annexin V Alexa Fluor 647 conjugate is excitable at 650 nm with an emission peak at 665 nm.
The Annexin V APC conjugate is excitable at 650 nm with an emission peak at 660 nm.
The Annexin V Pacific Blue conjugate is excitable at 410 nm with an emission peak at 455 nm.
Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.
Specifications
Description
Annexin V Alexa Fluor 488 Ready Flow Conjugate
Kit Contents
2 dropper bottles, store at room temperature
Product Type
Ready Flow Conjugate
Content And Storage
Store at room temperature.
Flow Cytometer Laser Lines
Blue
Excitation/Emission
495/519
Color
Green
Shipping Condition
Room Temperature
Conjugate
Alexa Fluor 488
For Use With (Equipment)
Flow Cytometer
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.