Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Anode Carbon Composite Toe

Men’s 4.5 in. 100% Non-Metallic ESD Carbon Composite Toe 4x4 Hiker
Supplier: Carolina™ CA55260952E
This item is not returnable.
View return policy
Description
- Brown
- Bandit Potting Soil Leather
- Composite Safety Toe
- ESD
- 6 Inch
- Broad Toe
Specifications
Brown | |
9.5 | |
Composite |
Leather | |
Rubber | |
2E |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction