Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Anode Carbon Composite Toe
SDP

Men’s 4.5 in. 100% Non-Metallic ESD Carbon Composite Toe 4x4 Hiker

Supplier:  Carolina™ CA5526115D

 View more versions of this product

Catalog No. 19-154-148

This item is not returnable. View return policy

Description

Description

  • Brown
  • Bandit Potting Soil Leather
  • Composite Safety Toe
  • ESD
  • 6 Inch
  • Broad Toe
Specifications

Specifications

Brown
11.5
Composite
Leather
Rubber
D
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.