Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anpirtoline hydrochloride, Tocris Bioscience™

Catalog No. p-4783258 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent 5-HT1B agonist; also 5-HT3 antagonist

Highly potent 5-HT1B receptor agonist (Ki values are 28, 150 and 1490 nM at 5-HT1B, 5-HT1A and 5-HT2 receptors respectively). Decreases central serotonin synthesis and attenuates aggressive behavior in vivo. Also acts as an antagonist at 5-HT3 receptors (Ki = 29.5 nM) and is brain penetrant.

Chemical Identifiers

CAS 99201-87-3
Molecular Formula C10H14Cl2N2S
Molecular Weight (g/mol) 265.196
InChI Key GRXDJABVNGUGCW-UHFFFAOYSA-N
Synonym anpirtoline hydrochloride, 2-chloro-6-piperidin-4-ylthio pyridine hydrochloride, 2-chloro-6-piperidin-4-ylsulfanylpyridine hydrochloride, pyridine, 2-chloro-6-4-piperidinylthio-, monohydrochloride, 2-chloro-6-piperidin-4-ylsulfanyl 2,6-14c2 pyridine hydrochloride 1:1, 2-chloro-6-piperidin-4-ylsulfanyl pyridine, hydrochloride, 2-chloro-6-4-piperidinylthio pyridine hydrochloride, 2-chloro-6-piperidin-4-ylsulfanyl pyridine hydrochloride
PubChem CID 6917943
IUPAC Name 2-chloro-6-piperidin-4-ylsulfanylpyridine;hydrochloride
SMILES C1CNCCC1SC2=NC(=CC=C2)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 265.2
Percent Purity >99%
Chemical Name or Material Anpirtoline hydrochloride
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.