Learn More
TARGETMOL CHEMICALS INC ANR 94 5MG
Supplier:  TARGETMOL CHEMICALS INC T22031
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ANR94 is a potent and selective adenosine A 2A receptor (AA 2A R) antagonist with an K i of 46 nM for hAA 2A R. ANR94 has the potential for Parkinson's disease research [1] [2]. purity: 98%
Catalog No. 50-258-0984
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.