Learn More
Ansell Encore Acclaim Latex Sterile Gloves
Catalog No. 19166675 Shop All Ansell Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy
Latex, powder free surgical glove with a comfortable, natural fit and feel.
Featuring Ansell formers and glove formulation, this glove's combination of fit and comfort together with optimized tensile strength, puncture and tear resistance properties enables healthcare workers to perform at their very best.
  • Proprietary formulation delivers a soft and sensitive yet robust surgical glove
  • Unique anti-slip, smooth external surface treatment for exceptional instrument handling and grip
  • DERMASHIELD Technology™ internal coating facilitates dry and damp donning as well as intra-operative changes
  • Tested for use with chemotherapy drugs
Specifications
Certifications/Compliance
ASTM D 1076, ISO9001, ISO13485, ASTM D 3577, Type I
Color
Cream
Exterior Finish
No Texture
Material
Latex
Powdered
Powder Free
Size
5.5
Sterility
Sterile
Thickness (English)
9 mil
Length (English)
11.7 in.
Length (Metric)
29.7 cm