Learn More
PIP™ Anser™ Semi-Rimless Safety Glasses
Semi-Rimless Safety Glasses feature wraparound temples with rubberized tips gently grip the wearer's head.
Supplier: PIP™ 250AN10111
Description
- Lightweight, semi-rimless design provides all day user comfort
- Sleek, sporty style with wraparound lens protection
- Bayonet temples with flexible rubber tips offer a comfortable, secure fit
- Soft rubber nose pads help keep the glasses in place
- Supplied with an adjustable neck cord
- 9 Base curve lens
- Meets ANSI Z87.1+
Specifications
Scratch-resistant | |
Black | |
Polycarbonate | |
99.99% UVA and UVB Light Absorption |
Clear | |
Wraparound Temples with Rubber Tips | |
Polycarbonate/PVC | |
Semi-rimless |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Warning - Prop 65 - Reproductive Harm