Learn More
Medchemexpress LLC HY-U00096 5mg Medchemexpress, Ansofaxine hydrochloride CAS:916918-84-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU000965MG
Encompass_Preferred
Medchemexpress, HY-U00096 5mg Ansofaxine hydrochloride CAS:916918-84-8 Ansofaxine hydrochloride (LY03005; LPM570065) is a triple reuptake inhibitor; inhibits serotonin, dopamine and norepinephrine reuptake with IC50 values of 723, 491 and 763 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3187
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.