Learn More
Actin Mouse anti-All, Clone: C4, MilliporeSigma™

Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1501R
Description
This Anti-Actin Antibody, near a.a. 50-70, clone C4 is validated for use in ELISA, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), western blotting for the detection of Actin and has published in over 70 citations.Specifications
Actin | |
Monoclonal | |
Unconjugated | |
NP_001091.1 | |
Mouse | |
100 μg | |
All species | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
C4 | |
Protein G Purified in 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% NaN3. | |
ACTA1, MPFD, ASMA, NEM1, CFTDM, ACTA, CFTD, NEM2, Alpha-actin-1, CFTD1, NEM3 | |
Purified chicken gizzard actin (Lessard, 1988). | |
Primary | |
2-8°C for up to 6 months. | |
IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only