Learn More
CD23 Mouse anti-Human, PE, Clone: EBVCS-5, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 341007
Description
The CD23 antibody, clone EBVCS-5, is derived from the hybridization of Sp2/0 mouse myeloma cells with spleen cells from BALB/c mice immunized with an in vitro–transformed EBV cell line.
The CD23 antibody recognizes a 45-kilodalton (kDa) type II membrane glycoprotein, which is a human B-lymphocyte differentiation antigen. The CD23 antigen is also known as the low affinity IgE receptor, Fc epsilon RII, and FcεRII.
Specifications
CD23 | |
Monoclonal | |
12.5μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
In vitro-transformed EBV cell line | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
EBVCS-5 | |
PE | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only.