Learn More
Dimethyl-Histone H3 (Arg26) Rabbit anti-Human, Tetrahymena, Vertebrates, Xenopus, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07215
Description
Anti-dimethyl-Histone H3 (Arg26) Antibody is a rabbit polyclonal antibody for detection of dimethyl-Histone H3 (Arg26) also known as H3R26me2, Histone H3 (di methyl R26) and has been validated in western blot, Mplex.Specifications
dimethyl-Histone H3 (Arg26) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl, with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Tetrahymena species, Vertebrates, Xenopus species |
Multiplex, Western Blot | |
Unconjugated | |
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t | |
KLH-conjugated synthetic peptide (ATKAA(di-Me)RKSAPA-C) corresponding to amino acids 21-31 of Human Histone H3. | |
100 μL | |
NP_003484.1 | |
2-8°C for 12 months |