Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC1 Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06720
Description
Use Anti-HDAC1 Antibody (Rabbit Polyclonal Antibody) validated in EA, immunocytochemistry, immunoprecipitation, western blotting to detect HDAC1 also known as histone deacetylase 1.Specifications
HDAC1 | |
Polyclonal | |
Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, containing 0.05% sodium azide. Frozen at −20°C. | |
Rabbit | |
Protein A chromatography | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
HDAC1; DKFZp686H12203; HD1; RPD3L1; GON-10; RPD3 | |
6X-His-tagged fragment representing amino acids 53-482 of mouse histone deacetylase 1 (HDAC1). | |
200 μg | |
NP_004955 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction