Learn More
HEY1, Mouse, Clone: 3G10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HEY1.
Supplier: Abnova Corporation H00023462M08
Description
This gene encodes a nuclear protein belonging to the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcriptional repressors. Expression of this gene is induced by the Notch and c-Jun signal transduction pathways. Two similar and redundant genes in mouse are required for embryonic cardiovascular development, and are also implicated in neurogenesis and somitogenesis. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: DYRSLGFRECLAEVARYLSIIEGLDASDPLRVRLVSHLNNYASQREAASGAHAGLGHIPWGTVFGHHPHIAHPLLLPQNGHGNAGTTASPTEPHHQGRLGSpecifications
HEY1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_012258 | |
HEY1 | |
HEY1 (NP_036390, 121 a.a. ∼ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
3G10 | |
Mouse monoclonal antibody raised against a partial recombinant HEY1. | |
HEY1 | |
BHLHb31/CHF2/HERP2/HESR1/HRT-1/MGC1274/OAF1 | |
Mouse | |
Affinity chromatography | |
RUO | |
23462 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.